siRNA Details
| Gene Symbol: DNAH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018897.2 | NP_061720.2 | 21 | 3414 | ||
| XM_011511487.2 | XP_011509789.1 | 22 | 3790 | |||
| XM_011511488.2 | XP_011509790.1 | 20 | 4464 | |||
| XM_011511489.2 | XP_011509791.1 | 20 | 3376 | |||
| XM_011511490.2 | XP_011509792.1 | 21 | 4584 | |||
| XM_011511491.2 | XP_011509793.1 | 21 | 4586 | |||
| XM_011511492.2 | XP_011509794.1 | 21 | 4586 | |||
| XM_011511493.2 | XP_011509795.1 | 21 | 4586 | |||
| XM_011511494.2 | XP_011509796.1 | 21 | 4585 | |||
| XM_011511495.2 | XP_011509797.1 | 21 | 4585 | |||
| XM_017004504.1 | XP_016859993.1 | 20 | 4310 | |||
| GenBank mRNA | AB023161.2 | - | 21 | 3432 | ||
| AF327442.1 | - | 21 | 3409 | |||

DNAH7
Genomic Map