siRNA Details
| Gene Symbol: ANKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054027.4 | NP_473368.1 | 2, 3 | 644 | ||
| XM_011514067.1 | XP_011512369.1 | 2, 3 | 607 | |||
| XM_017009644.1 | XP_016865133.1 | 2, 3 | 244 | |||
| GenBank mRNA | AB046801.1 | - | 2, 3 | 474 | ||
| AK222631.1 | - | 342 | ||||
| AK315012.1 | - | 2, 3 | 342 | |||
| AY358503.1 | - | 2, 3 | 395 | |||
| BC009835.2 | - | 2, 3 | 569 | 3927236 | ||
| BC014526.2 | - | 2, 3 | 369 | 3837372 | ||
| BU193060.1 | - | 2, 3 | 637 | |||

ANKH
Genomic Map