siRNA Details
| Gene Symbol: LMOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304418.1 | NP_001291347.1 | 3 | 1271 | ||
| NM_198271.4 | NP_938012.2 | 2 | 1365 | |||
| GenBank mRNA | AK057852.1 | - | 2 | 892 | ||
| AK300096.1 | - | 3 | 1244 | |||
| AL832033.1 | - | 2 | 1336 | |||
| AL832709.1 | - | 2 | 1370 | |||
| BC121019.2 | - | 2 | 1244 | 40117347 | ||

LMOD3
Genomic Map