siRNA Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304782.1 | NP_001291711.1 | 2 | 264 | ||
| NM_019604.3 | NP_062550.2 | 7 | 812 | |||
| XM_011542900.2 | XP_011541202.1 | 6 | 738 | |||
| GenBank mRNA | AB209830.1 | - | 2 | 173 | ||
| AF001622.1 | - | 7 | 763 | |||
| AK293470.1 | - | 2 | 193 | |||
| AK313578.1 | - | 7 | 810 | |||
| BC070266.1 | - | 7 | 812 | 30418882 | ||
| DC305734.1 | - | 2 | 264 | |||

CRTAM
Genomic Map