siRNA Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019604.3 | NP_062550.2 | 3 | 360 | ||
| GenBank mRNA | AF001622.1 | - | 3 | 311 | ||
| AK313578.1 | - | 3 | 358 | |||
| BC070266.1 | - | 3 | 360 | 30418882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRTAM| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019604.3 | NP_062550.2 | 3 | 360 | ||
| GenBank mRNA | AF001622.1 | - | 3 | 311 | ||
| AK313578.1 | - | 3 | 358 | |||
| BC070266.1 | - | 3 | 360 | 30418882 | ||