siRNA Details
| Gene Symbol: CPXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184699.1 | NP_001171628.1 | 10 | 1369 | ||
| NM_019609.4 | NP_062555.1 | 10 | 1369 | |||
| GenBank mRNA | AK027661.1 | - | 10 | 1305 | ||
| AK075508.1 | - | 6 | 581 | |||
| AK075527.1 | - | 10 | 1344 | |||
| AK303142.1 | - | 7 | 848 | |||
| AK315027.1 | - | 10 | 1356 | |||
| AY358956.1 | - | 10 | 1325 | |||
| BC063430.1 | - | 10 | 1346 | 5239864 | ||

CPXM1
Genomic Map