siRNA Details
| Gene Symbol: BEX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080425.3 | NP_001073894.1 | 3 | 473 | ||
| NM_001127688.2 | NP_001121160.1 | 2 | 390 | |||
| GenBank mRNA | AK000959.1 | - | 3 | 428 | ||
| AL544124.3 | - | 2 | 366 | |||
| AV726729.1 | - | 3 | 473 | |||
| AY833563.1 | - | 3 | 428 | |||

BEX4
Genomic Map