siRNA Details
| Gene Symbol: ANKRD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019644.3 | NP_062618.2 | 3 | 544 | ||
| GenBank mRNA | AK302407.1 | - | 3 | 498 | ||
| BC007020.2 | - | 1 | 78 | |||
| BC032799.1 | - | 3 | 305 | 5170634 | ||
| BP369869.1 | - | 3 | 544 | |||
| D78334.1 | - | 3 | 300 | |||

ANKRD7
Genomic Map