siRNA Details
| Gene Symbol: DNAJC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021800.2 | NP_068572.1 | 1, 2 | 246 | ||
| NM_201262.1 | NP_957714.1 | 1, 2 | 246 | |||
| GenBank mRNA | AF176012.1 | - | 1, 2 | 231 | ||
| AF176013.1 | - | 1, 2 | 163 | |||
| AK289537.1 | - | 1, 2 | 196 | |||
| BC017018.1 | - | 1, 2 | 182 | 3884093 | ||
| CB121724.1 | - | 1, 2 | 248 | |||
| CR457184.1 | - | 1, 2 | 78 | |||

DNAJC12
Genomic Map