siRNA Details
| Gene Symbol: DIABLO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278302.1 | NP_001265231.1 | 3 | 735 | ||
| NM_001278303.1 | NP_001265232.1 | 3 | 763 | |||
| NM_001278304.1 | NP_001265233.1 | 3 | 1365 | |||
| NM_001278342.1 | NP_001265271.1 | 3 | 764 | |||
| NM_019887.5 | NP_063940.1 | 5 | 1160 | |||
| NM_138930.3 | NP_620308.1 | 4 | 867 | |||
| GenBank mRNA | AF262240.1 | - | 4 | 361 | ||
| AF298770.1 | - | 3 | 183 | |||
| AK001399.1 | - | 3 | 1369 | |||
| AK024768.1 | - | 4 | 369 | |||
| AK057778.1 | - | 3 | 237 | |||
| AK222664.1 | - | 4 | 358 | |||
| AK315629.1 | - | 4 | 380 | |||
| AY313210.1 | - | 3 | 210 | |||
| BC004417.1 | - | 2 | 175 | |||
| BC011909.2 | - | 4 | 365 | 4137792 | ||
| BC046209.1 | - | 5 | 1082 | |||
| BC095468.1 | - | 4 | 351 | 30557745 | ||
| BF313712.1 | - | 3 | 237 | |||
| BF724951.1 | - | 3 | 231 | |||
| BM746689.1 | - | 3 | 206 | |||
| CF128595.1 | - | 4 | 351 | |||
| JX679472.1 | - | 2 | 51 | |||
| KU178725.1 | - | 4 | 342 | |||
| KU178726.1 | - | 3 | 210 | |||

DIABLO
Genomic Map