siRNA Details
| Gene Symbol: INPP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318502.1 | NP_001305431.1 | 8, 9 | 2074 | ||
| NM_019892.5 | NP_063945.2 | 8, 9 | 2077 | |||
| XM_017014926.1 | XP_016870415.1 | 8, 9 | 2116 | |||
| GenBank mRNA | AF187891.1 | - | 8, 9 | 1778 | ||
| AL832846.1 | - | 8, 9 | 2075 | |||
| BC028032.1 | - | 8, 9 | 2059 | 5242186 | ||
| U45974.1 | - | 2, 3 | 291 | |||

INPP5E
Genomic Map