siRNA Details
| Gene Symbol: ACKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001296.4 | NP_001287.2 | 3 | 524 | ||
| GenBank mRNA | AK313561.1 | - | 3 | 521 | ||
| BC008816.2 | - | 3 | 512 | 3959017 | ||
| BC011631.2 | - | 3 | 512 | 3958671 | ||
| BC020558.1 | - | 2 | 443 | 3451472 | ||
| BC101629.1 | - | 1 | 376 | 8069135 | ||
| BC112045.1 | - | 1 | 376 | 8327513 | ||
| BE901130.1 | - | 4 | 514 | |||
| BT006800.1 | - | 1 | 345 | |||
| U94888.1 | - | 1 | 360 | |||
| Y12815.1 | - | 1 | 357 | |||

ACKR2
Genomic Map