siRNA Details
| Gene Symbol: SAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142648.1 | NP_001136120.1 | 4 | 291 | ||
| NM_020150.4 | NP_064535.1 | 3 | 221 | |||
| GenBank mRNA | AF087897.1 | - | 3 | 188 | ||
| AF217959.1 | - | 3 | 188 | |||
| AF261717.1 | - | 3 | 188 | |||
| AF274026.1 | - | 3 | 187 | |||
| AF318066.1 | - | 2 | 88 | |||
| AK312469.1 | - | 3 | 176 | |||
| AL136724.1 | - | 3 | 204 | |||
| AY008268.1 | - | 3 | 212 | |||
| BC003658.2 | - | 3 | 186 | 3546972 | ||
| BG327663.1 | - | 4 | 260 | |||
| CR533492.1 | - | 2 | 88 | |||

SAR1A
Genomic Map