siRNA Details
| Gene Symbol: FMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305424.1 | NP_001292353.1 | 10 | 4467 | ||
| NM_020066.4 | NP_064450.3 | 9 | 4455 | |||
| XM_017001837.1 | XP_016857326.1 | 10 | 4490 | |||
| XM_017001839.1 | XP_016857328.1 | 10 | 3307 | |||
| XM_017001840.1 | XP_016857329.1 | 9 | 2707 | |||
| XM_017001841.1 | XP_016857330.1 | 9 | 3165 | |||
| XM_017001842.1 | XP_016857331.1 | 8 | 2386 | |||
| XM_017001843.1 | XP_016857332.1 | 7 | 2374 | |||
| GenBank mRNA | AB209153.1 | - | 10 | 3060 | ||
| AK297716.1 | - | 4 | 225 | |||
| AK297755.1 | - | 4 | 471 | |||
| AK298141.1 | - | 2 | 290 | |||
| DN990332.1 | - | 5 | 459 | |||

FMN2
Genomic Map