siRNA Details
| Gene Symbol: LGALS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020129.2 | NP_064514.1 | 3 | 399 | ||
| NM_203471.1 | NP_982297.1 | 4 | 704 | |||
| GenBank mRNA | AF123762.1 | - | 2 | 176 | ||
| AF267852.1 | - | 3 | 399 | |||
| AF367430.1 | - | 3 | 399 | |||
| AF367431.1 | - | 3 | 399 | |||
| AF367432.1 | - | 3 | 399 | |||
| AK312040.1 | - | 3 | 238 | |||
| BC022257.1 | - | 3 | 255 | 4770140 | ||
| BG434228.1 | - | 4 | 530 | |||
| FJ613341.1 | - | 3 | 203 | |||
| FJ613342.1 | - | 3 | 203 | |||

LGALS14
Genomic Map