siRNA Details
| Gene Symbol: LGALS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020129.2 | NP_064514.1 | 3, 4 | 526 | ||
| NM_203471.1 | NP_982297.1 | 4, 5 | 831 | |||
| GenBank mRNA | AF123762.1 | - | 2, 3 | 303 | ||
| AF267852.1 | - | 3, 4 | 526 | |||
| AF367430.1 | - | 526 | ||||
| AK312040.1 | - | 3, 4 | 365 | |||
| BC022257.1 | - | 3, 4 | 382 | 4770140 | ||
| FJ613341.1 | - | 3, 4 | 330 | |||
| FJ613342.1 | - | 3, 4 | 330 | |||

LGALS14
Genomic Map