siRNA Details
| Gene Symbol: CNGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286130.1 | NP_001273059.1 | 28 | 2914 | ||
| NM_001297.4 | NP_001288.3 | 28 | 2932 | |||
| XM_011522870.2 | XP_011521172.1 | 16 | 2201 | |||
| GenBank mRNA | AF042498.1 | - | 28 | 2919 | ||
| U58837.1 | - | 28 | 2924 | |||

CNGB1
Genomic Map