siRNA Details
| Gene Symbol: PNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020143.2 | NP_064528.1 | 2 | 306 | ||
| GenBank mRNA | AF164799.1 | - | 2 | 264 | ||
| AF349314.1 | - | 2 | 286 | |||
| AK223118.1 | - | 2 | 234 | |||
| AK225641.1 | - | 2 | 230 | |||
| AK291715.1 | - | 2 | 294 | |||
| AK313204.1 | - | 2 | 254 | |||
| BC008304.1 | - | 2 | 306 | 3344225 | ||
| BC018152.1 | - | 2 | 101 | |||
| L48692.1 | - | 2 | 245 | |||

PNO1
Genomic Map