siRNA Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020147.3 | NP_064532.1 | 3 | 806 | ||
| GenBank mRNA | AK313472.1 | - | 3 | 804 | ||
| AL360202.1 | - | 3 | 765 | |||
| BC027857.1 | - | 3 | 787 | 5222191 | ||
| BC072414.1 | - | 3 | 794 | 6167441 | ||

THAP10
Genomic Map