siRNA Details
| Gene Symbol: STARD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020151.3 | NP_064536.2 | 2 | 885 | ||
| XM_011511499.1 | XP_011509801.1 | 2 | 502 | |||
| GenBank mRNA | AF270647.1 | - | 2 | 812 | ||
| AK023234.1 | - | 1 | 159 | |||
| AK054615.1 | - | 2 | 251 | |||
| AK291190.1 | - | 2 | 267 | |||
| AL542720.3 | - | 2 | 887 | |||
| BC003028.1 | - | 2 | 868 | |||
| BC007894.2 | - | 3 | 643 | 4134933 | ||
| BC008894.2 | - | 2 | 868 | 3833465 | ||
| BC009998.2 | - | 3 | 643 | 4128869 | ||
| BC012774.2 | - | 2 | 835 | 3842611 | ||
| BC012793.2 | - | 2 | 849 | 4080200 | ||
| BC013279.2 | - | 2 | 868 | 3830320 | ||
| BC014076.2 | - | 2 | 845 | 4127007 | ||
| BC014274.2 | - | 2 | 864 | 3677198 | ||
| BC032106.1 | - | 2 | 859 | 4643387 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_139308, BC017524, AF244543, AK199440, AK217776, and others.

STARD7
Genomic Map