siRNA Details
| Gene Symbol: C2orf83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162483.1 | NP_001155955.1 | 4 | 575 | ||
| NM_020161.3 | NP_064546.3 | 3 | 480 | |||
| GenBank mRNA | AL359944.1 | - | 3 | 332 | ||
| BC131618.1 | - | 3 | 318 | 40116809 | ||
| CR749679.1 | - | 3 | 332 | |||

C2orf83
Genomic Map