siRNA Details
| Gene Symbol: FEM1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020177.2 | NP_064562.1 | 3 | 1537 | ||
| XM_005272035.4 | XP_005272092.1 | 3 | 1539 | |||
| XM_017009647.1 | XP_016865136.1 | 4 | 1623 | |||
| GenBank mRNA | AB058688.1 | - | 2 | 1065 | ||
| AF391093.1 | - | 2 | 975 | |||
| AK025265.1 | - | 1 | 35 | |||
| AK315803.1 | - | 3 | 1315 | |||
| AL365409.1 | - | 1 | 77 | |||
| AL831817.1 | - | 1 | 142 | |||
| AY249188.1 | - | 2 | 975 | |||
| BC028369.2 | - | 3 | 1539 | 4826273 | ||

FEM1C
Genomic Map