siRNA Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006109.1 | NP_001006109.1 | 4 | 689 | ||
| NM_020187.2 | NP_064572.2 | 4 | 516 | |||
| XM_005247636.3 | XP_005247693.1 | 4 | 701 | |||
| XM_017006877.1 | XP_016862366.1 | 4 | 533 | |||
| GenBank mRNA | AF201934.1 | - | 4 | 529 | ||
| AK025883.1 | - | 3 | 361 | |||
| AK309891.1 | - | 4 | 513 | |||
| BC009993.2 | - | 4 | 596 | 4130099 | ||
| BC010125.2 | - | 4 | 479 | 3945258 | ||
| BC050686.1 | - | 4 | 689 | 6155077 | ||
| BC088363.1 | - | 4 | 513 | 30398247 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173737, AK010550, AK018400, BC024401, BC064070.

HMCES
Genomic Map