siRNA Details
| Gene Symbol: C5orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020199.2 | NP_064584.1 | 3 | 868 | ||
| GenBank mRNA | AF226055.1 | - | 3 | 941 | ||
| AK074963.1 | - | 3 | 780 | |||
| AK298515.1 | - | 4 | 677 | |||
| AK315362.1 | - | 3 | 766 | |||
| AY157581.1 | - | 3 | 777 | |||
| BC020875.1 | - | 3 | 759 | 4610607 | ||

C5orf15
Genomic Map