siRNA Details
| Gene Symbol: OTUD7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020205.3 | NP_064590.2 | 7 | 1177 | ||
| XM_011509782.1 | XP_011508084.1 | 7 | 1174 | |||
| XM_011509783.2 | XP_011508085.1 | 7 | 1404 | |||
| XM_011509784.2 | XP_011508086.1 | 7 | 968 | |||
| XM_011509785.2 | XP_011508087.1 | 7 | 4601 | |||
| XM_011509786.1 | XP_011508088.1 | 7 | 1040 | |||
| XM_011509787.1 | XP_011508089.1 | 7 | 783 | |||
| XM_011509788.1 | XP_011508090.1 | 6 | 892 | |||
| XM_017001850.1 | XP_016857339.1 | 7 | 1040 | |||
| XM_017001851.1 | XP_016857340.1 | 6 | 1026 | |||
| GenBank mRNA | AJ293573.1 | - | 7 | 1020 | ||
| AK299790.1 | - | 7 | 1163 | |||
| BC020622.1 | - | 7 | 963 | |||
| BC072681.1 | - | 7 | 1114 | 6170513 | ||

OTUD7B
Genomic Map