siRNA Details
| Gene Symbol: ADPRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020233.4 | NP_064618.3 | 2 | 466 | ||
| XM_005256738.2 | XP_005256795.1 | 2 | 466 | |||
| GenBank mRNA | AB073393.1 | - | 2 | 437 | ||
| AF168715.1 | - | 1 | 430 | |||
| AK292629.1 | - | 2 | 432 | |||
| BC001294.2 | - | 1 | 421 | 3452502 | ||
| BC070155.1 | - | 2 | 461 | 4656998 | ||
| DA305828.1 | - | 2 | 466 | |||
| KF880964.1 | - | 1 | 391 | |||

ADPRM
Genomic Map