siRNA Details
| Gene Symbol: CNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286055.1 | NP_001272984.1 | 3 | 539 | ||
| NM_001286056.1 | NP_001272985.1 | 3 | 329 | |||
| NM_001839.4 | NP_001830.1 | 3 | 539 | |||
| XM_017000245.1 | XP_016855734.1 | 5 | 2951 | |||
| GenBank mRNA | AK294141.1 | - | 3 | 290 | ||
| AK298145.1 | - | 3 | 459 | |||
| AK312370.1 | - | 3 | 426 | |||
| AK316398.1 | - | 3 | 305 | |||
| BC025372.2 | - | 3 | 423 | 3505668 | ||
| BU149686.1 | - | 3 | 475 | |||
| CR541848.1 | - | 3 | 210 | |||
| DC422152.1 | - | 3 | 495 | |||
| DR157009.1 | - | 3 | 462 | |||
| S80562.1 | - | 3 | 293 | |||

CNN3
Genomic Map