siRNA Details
Gene Symbol: TOMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020243.4 | NP_064628.1 | 3 | 323 | ||
GenBank mRNA | AB040119.1 | - | 3 | 305 | ||
AB041906.1 | - | 3 | 306 | |||
AF163256.1 | - | 3 | 306 | |||
AK024731.1 | - | 3 | 316 | |||
AK223021.1 | - | 3 | 321 | |||
AK312076.1 | - | 3 | 297 | |||
BC009363.2 | - | 3 | 298 | 4302531 | ||
CB142641.1 | - | 3 | 323 | |||
CR456475.1 | - | 3 | 298 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
