siRNA Details
| Gene Symbol: CNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286055.1 | NP_001272984.1 | 6 | 1064 | ||
| NM_001286056.1 | NP_001272985.1 | 7 | 992 | |||
| NM_001839.4 | NP_001830.1 | 7 | 1202 | |||
| XM_017000245.1 | XP_016855734.1 | 9 | 3614 | |||
| GenBank mRNA | AK294141.1 | - | 7 | 953 | ||
| AK298145.1 | - | 6 | 984 | |||
| AK312370.1 | - | 7 | 1089 | |||
| AK316398.1 | - | 7 | 968 | |||
| BC025372.2 | - | 7 | 1086 | 3505668 | ||
| CR541848.1 | - | 7 | 873 | |||
| S80562.1 | - | 7 | 956 | |||

CNN3
Genomic Map