siRNA Details
| Gene Symbol: SDHAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020186.2 | NP_064571.1 | 2 | 1413 | ||
| GenBank mRNA | AF201933.1 | - | 2 | 302 | ||
| BC020621.1 | - | 2 | 295 | 4766494 | ||
| BC022858.1 | - | 2 | 303 | 4132451 | ||
| BC028409.1 | - | 2 | 1421 | |||

SDHAF3
Genomic Map