siRNA Details
| Gene Symbol: COQ9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020312.3 | NP_064708.1 | 5 | 658 | ||
| GenBank mRNA | AF161444.1 | - | 5 | 592 | ||
| AK290627.1 | - | 5 | 588 | |||
| AK293585.1 | - | 4 | 876 | |||
| AK295793.1 | - | 5 | 601 | |||
| AK303527.1 | - | 5 | 561 | |||
| BC001478.2 | - | 3 | 233 | |||
| BC029341.1 | - | 5 | 592 | |||
| BC054340.1 | - | 5 | 592 | 6514554 | ||
| BC064946.1 | - | 5 | 593 | 6023461 | ||

COQ9
Genomic Map