siRNA Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 759 | ||
| NM_001160258.1 | NP_001153730.1 | 4 | 797 | |||
| NM_001160259.1 | NP_001153731.1 | 2 | 672 | |||
| NM_016083.4 | NP_057167.2 | 2 | 616 | |||
| NM_033181.3 | NP_149421.2 | 2 | 271 | |||
| XM_006715330.3 | XP_006715393.1 | 2 | 563 | |||
| XM_011535424.1 | XP_011533726.1 | 3 | 1103 | |||
| XM_011535425.1 | XP_011533727.1 | 4 | 1141 | |||
| XM_011535426.1 | XP_011533728.1 | 5 | 1299 | |||
| XM_011535428.1 | XP_011533730.1 | 3 | 950 | |||
| XM_017010238.1 | XP_016865727.1 | 4 | 3927 | |||
| XM_017010239.1 | XP_016865728.1 | 3 | 2250 | |||
| XM_017010240.1 | XP_016865729.1 | 2 | 610 | |||
| GenBank mRNA | AF107262.1 | - | 1 | 316 | ||
| AK313908.1 | - | 2 | 437 | |||
| AY766182.1 | - | 2 | 208 | |||
| AY995204.1 | - | 1 | 307 | |||
| BC074811.2 | - | 1 | 345 | 30915316 | ||
| BC074812.2 | - | 1 | 345 | 30915532 | ||
| BC100968.1 | - | 1 | 307 | 40004970 | ||
| BC100969.1 | - | 1 | 307 | 40004975 | ||
| BC100970.1 | - | 1 | 307 | 40004978 | ||
| BC100971.1 | - | 1 | 307 | 40004979 | ||
| DQ067455.1 | - | 1 | 307 | |||
| KC292264.1 | - | 2 | 271 | |||
| X54937.1 | - | 2 | 455 | |||
| X81120.1 | - | 1 | 398 | |||
| X81121.1 | - | 2 | 140 | |||
More Information
Set Membership: |
|

CNR1
Genomic Map