siRNA Details
| Gene Symbol: CHRNA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303034.1 | NP_001289963.1 | 5 | 1092 | ||
| NM_001303035.1 | NP_001289964.1 | 5 | 1025 | |||
| NM_020402.3 | NP_065135.2 | 5 | 1030 | |||
| XM_011520234.1 | XP_011518536.1 | 4 | 4226 | |||
| GenBank mRNA | AF199235.2 | - | 5 | 1030 | ||
| AJ278118.1 | - | 5 | 1000 | |||
| AJ295237.1 | - | 5 | 1000 | |||
| BC136250.1 | - | 5 | 1008 | |||
| BC143303.1 | - | 5 | 1075 | |||

CHRNA10
Genomic Map