siRNA Details
| Gene Symbol: AVEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020371.2 | NP_065104.1 | 2 | 418 | ||
| XM_011521818.2 | XP_011520120.1 | 3 | 601 | |||
| XM_011521819.2 | XP_011520121.1 | 2 | 644 | |||
| XM_011521820.1 | XP_011520122.1 | 2 | 119 | |||
| GenBank mRNA | AF283508.1 | - | 2 | 414 | ||
| BC010488.1 | - | 2 | 412 | 2900586 | ||
| BC063533.1 | - | 2 | 402 | 4425400 | ||
| BU855198.1 | - | 492 | ||||

AVEN
Genomic Map