siRNA Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000314.6 | NP_000305.3 | 9 | 4421 | ||
| NM_001304717.2 | NP_001291646.2 | 9 | 4421 | |||
| NM_001304718.1 | NP_001291647.1 | 9 | 4535 | |||
| GenBank mRNA | AK024986.1 | - | 1 | 957 | ||

PTEN
Genomic Map