siRNA Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020387.3 | NP_065120.2 | 3 | 620 | ||
| NR_133653.1 | 2 | 424 | ||||
| GenBank mRNA | AF083124.1 | - | 3 | 581 | ||
| AF274025.1 | - | 3 | 599 | |||
| BC004416.1 | - | 1 | 110 | |||
| BC009831.2 | - | 3 | 594 | 3926839 | ||
| BC033322.1 | - | 3 | 584 | 4775725 | ||
| BP273992.1 | - | 2 | 409 | |||

RAB25
Genomic Map