siRNA Details
| Gene Symbol: RNPEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018226.4 | NP_060696.4 | 11 | 1900 | ||
| XM_005247036.4 | XP_005247093.1 | 11 | 1928 | |||
| GenBank mRNA | AF300795.1 | - | 11 | 1606 | ||
| AK001668.1 | - | 7 | 864 | |||
| AK130185.1 | - | 4 | 468 | |||
| AK130805.1 | - | 4 | 2834 | |||
| AL512754.1 | - | 4 | 485 | |||
| BC017301.2 | - | 8 | 1052 | |||
| BC067258.1 | - | 11 | 1475 | 6183046 | ||
| BC082975.1 | - | 11 | 1800 | 6379537 | ||
| BC099906.1 | - | 9 | 2013 | 5311883 | ||

RNPEPL1
Genomic Map