siRNA Details
| Gene Symbol: TMEM63C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020431.3 | NP_065164.2 | 21 | 2113 | ||
| GenBank mRNA | AK125159.1 | - | 21 | 2045 | ||
| AL157492.1 | - | 2 | 66 | |||
| BC136613.1 | - | 21 | 1995 | 9020603 | ||
| BC136614.1 | - | 21 | 1992 | 9020604 | ||

TMEM63C
Genomic Map