siRNA Details
| Gene Symbol: PELI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021255.2 | NP_067078.1 | 1, 2 | 369 | ||
| XM_005267890.4 | XP_005267947.1 | 1, 2 | 414 | |||
| GenBank mRNA | AF302502.1 | - | 1, 2 | 259 | ||
| AK315727.1 | - | 1, 2 | 331 | |||
| BC009476.2 | - | 1, 2 | 369 | 3942712 | ||

PELI2
Genomic Map