siRNA Details
| Gene Symbol: MCOLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020533.2 | NP_065394.1 | 4 | 703 | ||
| GenBank mRNA | AF249319.1 | - | 4 | 646 | ||
| AF287269.1 | - | 4 | 687 | |||
| AJ293659.1 | - | 4 | 642 | |||
| AJ293970.1 | - | 4 | 642 | |||
| AK026102.1 | - | 4 | 657 | |||
| AK222673.1 | - | 4 | 682 | |||
| AK294330.1 | - | 4 | 668 | |||
| BC005149.2 | - | 4 | 685 | 3507836 | ||

MCOLN1
Genomic Map