siRNA Details
| Gene Symbol: THAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020457.2 | NP_065190.2 | 1 | 318 | ||
| GenBank mRNA | AK289367.1 | - | 1 | 294 | ||
| BC012182.1 | - | 1 | 280 | 4554554 | ||
| BI261822.1 | - | 1 | 228 | |||
| BQ898449.1 | - | 318 | ||||
| L10378.1 | - | 1 | 155 | |||
| L10379.1 | - | 1 | 155 | |||

THAP11
Genomic Map