siRNA Details
| Gene Symbol: LYRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020466.4 | NP_065199.1 | 2 | 171 | ||
| NR_028493.1 | 2 | 313 | ||||
| NR_028494.1 | 2 | 313 | ||||
| NR_028495.1 | 2 | 181 | ||||
| GenBank mRNA | AK024783.1 | - | 2 | 169 | ||
| AK094994.1 | - | 2 | 222 | |||
| AK311151.1 | - | 2 | 313 | |||
| AK311180.1 | - | 2 | 230 | |||
| AK311275.1 | - | 2 | 166 | |||
| AL122090.1 | - | 2 | 154 | |||
| AV710669.1 | - | 2 | 171 | |||
| BC009782.1 | - | 2 | 165 | 4092710 | ||
| BC104668.1 | - | 2 | 154 | 6717725 | ||
| BP280644.1 | - | 2 | 171 | |||
| DA160441.1 | - | 2 | 313 | |||

LYRM2
Genomic Map