siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 3 | 325 | ||
| GenBank mRNA | AF294429.1 | - | 2 | 217 | ||
| AK294921.1 | - | 3 | 324 | |||
| AK304614.1 | - | 3 | 327 | |||
| AK310427.1 | - | 4 | 1134 | |||
| AK311485.1 | - | 3 | 327 | |||
| BC015942.1 | - | 3 | 325 | |||
| BC034633.1 | - | 3 | 279 | |||
| BC136261.1 | - | 3 | 273 | 9020248 | ||
| BC143547.1 | - | 3 | 273 | 9052058 | ||
| BC151233.1 | - | 4 | 419 | |||
| CR749504.1 | - | 5 | 1204 | |||

CLK4
Genomic Map