siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 6 | 743 | ||
| GenBank mRNA | AF294429.1 | - | 5 | 635 | ||
| AK311485.1 | - | 5 | 654 | |||
| BC015942.1 | - | 6 | 743 | |||
| BC136261.1 | - | 6 | 691 | 9020248 | ||
| BC143547.1 | - | 6 | 691 | 9052058 | ||
| BC151233.1 | - | 6 | 1957 | |||
| BX647323.1 | - | 2 | 600 | |||
| CR749504.1 | - | 8 | 1622 | |||

CLK4
Genomic Map