siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 12 | 1355 | ||
| GenBank mRNA | AF294429.1 | - | 11 | 1247 | ||
| AK311485.1 | - | 12 | 1307 | |||
| BC015942.1 | - | 12 | 1355 | |||
| BC136261.1 | - | 12 | 1303 | 9020248 | ||
| BC151233.1 | - | 12 | 2569 | |||
| BX647323.1 | - | 6 | 2497 | |||
| CR749504.1 | - | 14 | 2234 | |||

CLK4
Genomic Map