siRNA Details
| Gene Symbol: C3orf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291941.1 | NP_001278870.1 | 4, 5 | 382 | ||
| NM_001291942.1 | NP_001278871.1 | 3, 4 | 270 | |||
| NM_020685.4 | NP_065736.1 | 4, 5 | 489 | |||
| XM_017006931.1 | XP_016862420.1 | 4, 5 | 865 | |||
| GenBank mRNA | AF236158.1 | - | 4, 5 | 319 | ||
| AK313915.1 | - | 4, 5 | 489 | |||
| BC017772.2 | - | 3, 4 | 299 | 4307022 | ||
| BU568674.1 | - | 224 | ||||
| DA783189.1 | - | 4, 5 | 382 | |||

C3orf14
Genomic Map