siRNA Details
| Gene Symbol: WDR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024100.3 | NP_077005.2 | 7 | 844 | ||
| XM_017027027.1 | XP_016882516.1 | 9 | 1370 | |||
| GenBank mRNA | BC000040.2 | - | 7 | 829 | 3507673 | |
| BC001648.2 | - | 7 | 824 | 2819452 | ||

WDR18
Genomic Map