siRNA Details
| Gene Symbol: KIAA1143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320334.1 | NP_001307263.1 | 1 | 176 | ||
| NM_020696.3 | NP_065747.1 | 1 | 176 | |||
| GenBank mRNA | AK289549.1 | - | 1 | 148 | ||
| BC008468.1 | - | 1 | 151 | 4277104 | ||
| BC016790.1 | - | 1 | 110 | 4102473 | ||
| BC070360.1 | - | 1 | 106 | |||
| CD049061.1 | - | 1 | 173 | |||
| DA183520.1 | - | 1 | 169 | |||
| DA192476.1 | - | 1 | 151 | |||
| DB238094.1 | - | 1 | 132 | |||

KIAA1143
Genomic Map