siRNA Details
| Gene Symbol: ERMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 1086 | ||
| NM_001304344.1 | NP_001291273.1 | 4 | 934 | |||
| NM_001304345.1 | NP_001291274.1 | 4 | 946 | |||
| NM_001304346.1 | NP_001291275.1 | 3 | 1059 | |||
| NM_020711.2 | NP_065762.1 | 3 | 1119 | |||
| GenBank mRNA | AB033015.1 | - | 3 | 861 | ||
| AK295051.1 | - | 2 | 794 | |||
| AK295844.1 | - | 3 | 935 | |||
| AK296477.1 | - | 4 | 1029 | |||
| AK296489.1 | - | 3 | 680 | |||
| BC026345.1 | - | 3 | 945 | 4798869 | ||
| DQ334271.1 | - | 3 | 942 | |||

ERMN
Genomic Map