siRNA Details
| Gene Symbol: ERMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 783 | ||
| NM_001304344.1 | NP_001291273.1 | 4 | 631 | |||
| NM_001304345.1 | NP_001291274.1 | 4 | 643 | |||
| NM_001304346.1 | NP_001291275.1 | 3 | 756 | |||
| NM_020711.2 | NP_065762.1 | 3 | 816 | |||
| GenBank mRNA | AB033015.1 | - | 3 | 558 | ||
| AK295051.1 | - | 2 | 491 | |||
| AK295844.1 | - | 3 | 632 | |||
| AK296477.1 | - | 4 | 726 | |||
| AK296489.1 | - | 3 | 377 | |||
| BC026345.1 | - | 3 | 642 | 4798869 | ||
| DQ334271.1 | - | 3 | 639 | |||

ERMN
Genomic Map